missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD300C Protein

A cDNA sequence encoding the CD300C was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11325-HIS-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15966858

  • £2978.71 / 1mg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

10871
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
CD300C
Human
His
CD300C protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol.
CD300C Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Insect
MGSSHHHHHH SSGLVPRGSH MGSGYFPLSH PMTVAGPVGG SLSVQCRYEK EHRTLNKFWC RPPQILRCDK IVETKGSAGK RNGRVSIRDS PANLSFTVTL ENLTEEDAGT YWCGVDTPWL RDFHDPIVEV EVSVFPAGTT TASSPQSSMG TSGPPTKLPV HTWPSVTRKD SPEPSPHPGSLFSNVR
Videos
SDS
Documents
Certificates
Special Offers

Special Offers