missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD27 Protein
A cDNA sequence encoding the CD27 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11318-HIS-INSECT-5ug
Additional Details : Weight : 0.01000kg
Specifications
939 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MGSSHHHHHH SSGLVPRGSH MGSHMTPAPK SCPERHYWAQ GKLCCQMCEP GTFLVKDCDQ HRKAAQCDPC IPGVSFSPDH HTRPHCESCR HCNSGLLVRN CTITANAECA CRNGWQCRDK ECTECDPLPN PSLTARSSQA LSPHPQPTHL PYVSEMLEAR TAGHMQTLAD FRQLPARTLS THWPPQRSLC SSDFIR | |
Greater than 90% as determined by SDS-PAGE. |
CD27 Protein | |
Research Use Only | |
CD27 | |
Human | |
His | |
The CD27 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |