missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD226 Protein
A cDNA sequence encoding the CD226 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11315-HIS-EC-1mg
Additional Details : Weight : 0.01000kg
Specifications
10666 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEEVLWHT SVPFAENMSL ECVYPSMGIL TQVEWFKIGT QQDSIAIFSP THGMVIRKPY AERVYFLNST MASNNMTLFF RNASEDDVGY YSCSLYTYPQ GTWQKVIQVV QSDSFEAAVP SNSHIVSEPG KNVTLTCQPQ MTWPVQAVRW EKIQPRQIDL LTYCNLVHGR NFTSKFPRQI VSNCSHGRWS VIVIPDVTVS DSGLYRCYLQ ASAGENETFV MRLTVAEGKT DNQYTLFVA | |
Greater than 90% as determined by SDS-PAGE. |
CD226 Protein | |
Research Use Only | |
CD226 | |
Human | |
His | |
CD226 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |