missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD1D Protein

A cDNA sequence encoding the CD1D was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11310-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956798

  • £155.00 / 5µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD1D Protein
Research Use Only
Recombinant Protein
Baculovirus
EVPQRLFPLR CLQISSFANS SWTRTDGLAW LGELQTHSWS NDSDTVRSLK PWSQGTFSDQ QWETLQHIFR VYRSSFTRDV KEFAKMLRLS YPLELQVSAG CEVHPGNASN NFFHVAFQGK DILSFQGTSW EPTQEAPLWV NLAIQVLNQD KWTRETVQWL LNGTCPQFVS GLLESGKSELKKQVKPKAWL SRGPSPGPGR LLLVCHVSGF YPKPVWVKWM RGEQEQQGTQ PGDILPNADE TWYLRATLDV VAGEAAGLSC RVKHSSLEGQ DIVLYWGGSY TSLEHHHHHH
Greater than 85.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CD1D protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers