missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD164L2 Protein
A cDNA sequence encoding the CD164L2 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Gene ID (Entrez) | 388611 |
---|---|
Name | CD164L2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CD164L2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15936788
|
enQuireBio™
QP11306-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926788
|
enQuireBio™
QP11306-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15916788
|
enQuireBio™
QP11306-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
388611 | |
Research Use Only | |
CD164L2 | |
Human | |
His | |
CD164L2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |
CD164L2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGKGARGF GRGALIRLNI WPAVQGACKQ LEVCEHCVEG DRARNLSSCM WEQCRPEEPG HCVAQSEVVK EGCSIYNRSE ACPAAHHHPT YEPKTVTTGS PPVPEAHSPG FDGAS | |
Greater than 90% as determined by SDS-PAGE. |