missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCNH Protein
A cDNA sequence encoding the CCNH was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11297-5ug
Additional Details : Weight : 0.01000kg
Specifications
902 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MYHNSSQKRH WTFSSEEQLA RLRADANRKF RCKAVANGKV LPNDPVFLEP HEEMTLCKYY EKRLLEFCSV FKPAMPRSVV GTACMYFKRF YLNNSVMEYH PRIIMLTCAF LACKVDEFNV SSPQFVGNLR ESPLGQEKAL EQILEYELLL IQQLNFHLIV HNPYRPFEGF LIDLKTRYPI LENPEILRKT ADDFLNRIAL TDAYLLYTPS QIALTAILSS ASRAGITMES YLSESLMLKE NRTCLSQLLD IMKSMRNLVK KYEPPRSEEV AVLKQKLERC HSAELALNVI TKKRKGYEDD DYVSKKSKHE EEEWTDDDLV ESL | |
Greater than 90% as determined by SDS-PAGE. |
CCNH Protein | |
Research Use Only | |
CCNH | |
Human | |
His | |
The CCNH solution contains 20mM Tris-HCl pH 8.0, 2mM DTT, 2mM EDTA, 0.1M NaCl and 30% glycerol. |