missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL4 / MIP1B Protein
Recombinant Protein
Brand: enQuireBio™ QP5256-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_002975.1 | |
CCL4 / MIP1B Protein | |
Research Use Only | |
CCL4 | |
Recombinant Protein | |
E. coli | |
APMGSDPPTA CCFSYTARKL PHNFVVDYYE TSSLCSQPAV VFQTKRGKQV CADPSESWVQ EYVYDLELN | |
Greater than 95% as determined by SDS-PAGE. |
388372 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human CCL4 / MIP1B induces Human PBMC chemotaxis (significanty different from control at Human CCL4 / MIP1B concentrations greater than 3.1 ng/ml). | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |