missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL24 / Eotaxin-2 / MPIF-2 Protein
A DNA sequence encoding the Homo sapiens (Human) CCL24 / Eotaxin-2 / MPIF-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8522-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002982.2 | |
CCL24 / Eotaxin-2 / MPIF-2 Protein | |
Research Use Only | |
CCL24 | |
Recombinant Protein | |
E. coli | |
VVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKAGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVAVK GPVQRYPGNQ TTC | |
Greater than 90% as determined by SDS-PAGE. |
6369 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |