missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL24 / Eotaxin-2 / MPIF-2 Protein
A DNA sequence encoding the Homo sapiens (Human) CCL24 / Eotaxin-2 / MPIF-2, was expressed in the hosts and tags indicated.
£145.46 - £1688.38
Specifications
Accession Number | NP_002982.2 |
---|---|
Gene ID (Entrez) | 6369 |
Name | CCL24 / Eotaxin-2 / MPIF-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16075091
|
enQuireBio™
QP8522-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015101
|
enQuireBio™
QP8522-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065091
|
enQuireBio™
QP8522-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095091
|
enQuireBio™
QP8522-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005101
|
enQuireBio™
QP8522-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085091
|
enQuireBio™
QP8522-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002982.2 | |
CCL24 / Eotaxin-2 / MPIF-2 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
6369 | |
Research Use Only | |
CCL24 | |
Recombinant Protein | |
E. coli | |
VVIPSPCCMF FVSKRIPENR VVSYQLSSRS TCLKAGVIFT TKKGQQFCGD PKQEWVQRYM KNLDAKQKKA SPRARAVAVK GPVQRYPGNQ TTC | |
Greater than 90% as determined by SDS-PAGE. |