missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCDC23 Protein
A cDNA sequence encoding the CCDC23 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11288-1mg
Additional Details : Weight : 0.01000kg
Specifications
CCDC23 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEFMDPPA RKEKTKVKES VSRVEKAKQK SAQQELKQRQ RAEIYALNRV MTELEQQQFD EFCKQMQPPG E | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CCDC23 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT. |