missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cathepsin B / CTSB Protein

A DNA sequence encoding the Homo sapiens (Human) Cathepsin B / CTSB, was expressed in the hosts and tags indicated.

£145.46 - £1688.38

Specifications

Name Cathepsin B / CTSB Protein
Product Type Recombinant Protein
Protein Tag GST
Host Species E. coli
Cross Reactivity Human
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16015301
View Documents
enQuireBio™
QP8562-EC-10UG
10 μg £145.46
10µg View alternative products
Save Up to 
»
Estimated Shipment
03-11-2020
Please to purchase this item. Need a web account? Register with us today!
16055301
View Documents
enQuireBio™
QP8562-EC-50UG
50 μg £214.03
50µg View alternative products
Save Up to 
»
Estimated Shipment
03-11-2020
Please to purchase this item. Need a web account? Register with us today!
16005301
View Documents
enQuireBio™
QP8562-EC-100UG
100 μg £369.88
100µg View alternative products
Save Up to 
»
Estimated Shipment
03-11-2020
Please to purchase this item. Need a web account? Register with us today!
16035301
View Documents
enQuireBio™
QP8562-EC-200UG
200 μg £613.01
200µg View alternative products
Save Up to 
»
Estimated Shipment
03-11-2020
Please to purchase this item. Need a web account? Register with us today!
16045301
View Documents
enQuireBio™
QP8562-EC-500UG
500 μg £1075.37
500µg View alternative products
Save Up to 
»
Estimated Shipment
03-11-2020
Please to purchase this item. Need a web account? Register with us today!
16025301
View Documents
enQuireBio™
QP8562-EC-1MG
1 mg £1688.38
1mg View alternative products
Save Up to 
»
Estimated Shipment
03-11-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Specifications

Cathepsin B / CTSB Protein
GST
Human
Greater than 90% as determined by SDS-PAGE.
NP_001899.1
Recombinant Protein
E. coli
ASFDAREQWP QCPTIKEIRD QGSCGSCWAF GAVEAISDRI CIHTNAHVSV EVSAEDLLTC CGSMCGDGCN GGYPAEAWNF WTRKGLVSGG LYESHVGCRP YSIPPCEHHV NGSRPPCTGE GDTPKCSKIC EPGYSPTYKQ DKHYGYNSYS VSNSEKDIMA EIYKNGPVEG AFSVYSDFLL YKSGVYQHVT GEMMGGHAIR ILGWGVENGT PYWLVANSWN TDWGDNGFFK ILRGQDHCGI ESEVVAGIPR TD
1508
CTSB