missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cathepsin B / CTSB Protein

A DNA sequence encoding the Homo sapiens (Human) Cathepsin B / CTSB, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8562-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16045301

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001899.1
Cathepsin B / CTSB Protein
Research Use Only
CTSB
Recombinant Protein
E. coli
ASFDAREQWP QCPTIKEIRD QGSCGSCWAF GAVEAISDRI CIHTNAHVSV EVSAEDLLTC CGSMCGDGCN GGYPAEAWNF WTRKGLVSGG LYESHVGCRP YSIPPCEHHV NGSRPPCTGE GDTPKCSKIC EPGYSPTYKQ DKHYGYNSYS VSNSEKDIMA EIYKNGPVEG AFSVYSDFLL YKSGVYQHVT GEMMGGHAIR ILGWGVENGT PYWLVANSWN TDWGDNGFFK ILRGQDHCGI ESEVVAGIPR TD
Greater than 90% as determined by SDS-PAGE.
1508
500 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers