missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cathepsin B / CTSB Protein
A DNA sequence encoding the Homo sapiens (Human) Cathepsin B / CTSB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8562-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001899.1 | |
Cathepsin B / CTSB Protein | |
Research Use Only | |
CTSB | |
Recombinant Protein | |
E. coli | |
ASFDAREQWP QCPTIKEIRD QGSCGSCWAF GAVEAISDRI CIHTNAHVSV EVSAEDLLTC CGSMCGDGCN GGYPAEAWNF WTRKGLVSGG LYESHVGCRP YSIPPCEHHV NGSRPPCTGE GDTPKCSKIC EPGYSPTYKQ DKHYGYNSYS VSNSEKDIMA EIYKNGPVEG AFSVYSDFLL YKSGVYQHVT GEMMGGHAIR ILGWGVENGT PYWLVANSWN TDWGDNGFFK ILRGQDHCGI ESEVVAGIPR TD | |
Greater than 90% as determined by SDS-PAGE. |
1508 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |