missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CASQ2 Protein
A cDNA sequence encoding the CASQ2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11281-25ug
Additional Details : Weight : 0.01000kg
Specifications
CASQ2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMEEG LNFPTYDGKD RVVSLSEKNF KQVLKKYDLL CLYYHEPVSS DKVTQKQFQL KEIVLELVAQ VLEHKAIGFV MVDAKKEAKL AKKLGFDEEG SLYILKGDRT IEFDGEFAAD VLVEFLLDLI EDPVEIISSK LEVQAFERIE DYIKLIGFFK SEDSEYYKAF EEAAEHFQPY IKFFATFDKG VAKKLSLKMN EVDFYEPFMD EPIAIPNKPY TEEELVEFVK EHQRPTLRRL RPEEMFETWE DDLNGIHIVA FAEKSDPDGY EFLEILKQVA RDNTDNPDLS ILWIDPDDFP LLVAYWEKTF KIDLFRPQIG VVNVTDADSV WMEIPDDDDL PTAEELEDWI EDVLSGKINT EDDDEDDDDD DNSDEEDNDD SDDDDDE | |
Greater than 95% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CASQ2 solution contains 20mM Tris-HCl pH 8.0, 0.1M NaCl, 1mM DTT and 20% glycerol. |