missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Caspase-14 / CASP14 Protein

A DNA sequence encoding the Homo sapiens (Human) Caspase-14 / CASP14, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5761-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15957559

  • £393.00 / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_036246.1
Caspase-14 / CASP14 Protein
Research Use Only
CASP14
Recombinant Protein
E. coli
KDSPQTIPTY TDALHVYSTV EGYIAYRHDQ KGSCFIQTLV DVFTKRKGHI LELLTEVTRR MAEAELVQEG KARKTNPEIQ STLRKRLY
Greater than 90% as determined by SDS-PAGE.
23581
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions