missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Caspase-14 / CASP14 Protein
A DNA sequence encoding the Homo sapiens (Human) Caspase-14 / CASP14, was expressed in the hosts and tags indicated.
£99.15 - £1780.00
Specifications
Accession Number | NP_036246.1 |
---|---|
Gene ID (Entrez) | 23581 |
Name | Caspase-14 / CASP14 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15967559
|
enQuireBio™
QP5761-EC-10UG |
10 μg |
£99.15
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15907569
|
enQuireBio™
QP5761-EC-50UG |
50 μg |
£246.00
50µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15957559
|
enQuireBio™
QP5761-EC-100UG |
100 μg |
£393.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15987559
|
enQuireBio™
QP5761-EC-200UG |
200 μg |
£635.00
200µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15997559
|
enQuireBio™
QP5761-EC-500UG |
500 μg |
£1095.00
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15977559
|
enQuireBio™
QP5761-EC-1MG |
1 mg |
£1780.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_036246.1 | |
Caspase-14 / CASP14 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
23581 | |
Research Use Only | |
CASP14 | |
Recombinant Protein | |
E. coli | |
KDSPQTIPTY TDALHVYSTV EGYIAYRHDQ KGSCFIQTLV DVFTKRKGHI LELLTEVTRR MAEAELVQEG KARKTNPEIQ STLRKRLY | |
Greater than 90% as determined by SDS-PAGE. |