missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CALM2 Protein
A cDNA sequence encoding the CALM2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11262-25ug
Additional Details : Weight : 0.01000kg
Specifications
CALM2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MADQLTEEQI AEFKEAFSLF DKDGDGTITT KELGTVMRSL GQNPTEAELQ DMINEVDADG NGTIDFPEFL TMMARKMKDT DSEEEIREAF RVFDKDGNGY ISAAELRHVM TNLGEKLTDE EVDEMIREAD IDGDGQVNYE EFVQMMTAK | |
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The CALM2 solution (1 mg/ml) contains 20mM Tris-HCl, pH 7.5. |