missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CALM2 Protein

A cDNA sequence encoding the CALM2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11262-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15966648

  • £3965.00 / 1mg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CALM2 Protein
Research Use Only
Recombinant Protein
E. coli
MADQLTEEQI AEFKEAFSLF DKDGDGTITT KELGTVMRSL GQNPTEAELQ DMINEVDADG NGTIDFPEFL TMMARKMKDT DSEEEIREAF RVFDKDGNGY ISAAELRHVM TNLGEKLTDE EVDEMIREAD IDGDGQVNYE EFVQMMTAK
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The CALM2 solution (1 mg/ml) contains 20mM Tris-HCl, pH 7.5.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers