missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CACNA2D1 Protein

A DNA sequence encoding the Homo sapiens (Human) CACNA2D1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5745-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927469

  • £635.00 / 200µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000713.2
CACNA2D1 Protein
Research Use Only
CACNA2D1
Recombinant Protein
Yeast
QPKPIGVGIP TINLRKRRPN IQNPKSQEPV TLDFLDAELE NDIKVEIRNK MIDGESGEKT FRTLVKSQDE RYIDKGNRTY TWTPVNGTDY SLALVLPTYS FYYIKAKLEE TITQARYSET LKPDNFEESG YTFIAPRDYC N
Greater than 90% as determined by SDS-PAGE.
781
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers