missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CABP7 Protein

A cDNA sequence encoding the CABP7 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11251-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15946618

  • £1320.00 / 50µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CABP7 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MPFHPVTAAL MYRGIYTVPN LLSEQRPVDI PEDELEEIRE AFKVFDRDGN GFISKQELGT AMRSLGYMPN EVELEVIIQR LDMDGDGQVD FEEFVTLLGP KLSTSGIPEK FHGTDFDTVF WKCDMQKLTV DELKRLLYDT FCEHLSMKDI ENIIMTEEES HLGTAEECPV DVETCSNQQI RQTCVRKS
Greater than 80.0% as determined by SDS-PAGE.
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CABP7 protein solution (0.5 mg/ml) containing Phosphate-buffered saline (pH 7.4).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers