missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CABP7 Protein
A cDNA sequence encoding the CABP7 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11251-1ug
Additional Details : Weight : 0.01000kg
Specifications
CABP7 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPFHPVTAAL MYRGIYTVPN LLSEQRPVDI PEDELEEIRE AFKVFDRDGN GFISKQELGT AMRSLGYMPN EVELEVIIQR LDMDGDGQVD FEEFVTLLGP KLSTSGIPEK FHGTDFDTVF WKCDMQKLTV DELKRLLYDT FCEHLSMKDI ENIIMTEEES HLGTAEECPV DVETCSNQQI RQTCVRKS | |
Greater than 80.0% as determined by SDS-PAGE. |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CABP7 protein solution (0.5 mg/ml) containing Phosphate-buffered saline (pH 7.4). |