missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CA3 Protein
A cDNA sequence encoding the CA3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11248-1mg
Additional Details : Weight : 0.01000kg
Specifications
761 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MAKEWGYASH NGPDHWHELF PNAKGENQSP IELHTKDIRH DPSLQPWSVS YDGGSAKTIL NNGKTCRVVF DDTYDRSMLR GGPLPGPYRL RQFHLHWGSS DDHGSEHTVD GVKYAAELHL VHWNPKYNTF KEALKQRDGI AVIGIFLKIG HENGEFQIFL DALDKIKTKG KEAPFTKFDP SCLFPACRDY WTYQGSFTTP PCEECIVWLL LKEPMTVSSD QMAKLRSLLS SAENEPPVPL VSNWRPPQPI NNRVVRASFK | |
Greater than 90% as determined by SDS-PAGE. |
CA3 Protein | |
Research Use Only | |
CA3 | |
Human | |
Untagged | |
The CA3 solution contains 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol. |