missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CA14 Protein
A cDNA sequence encoding the CA14 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11247-5ug
Additional Details : Weight : 0.01000kg
Specifications
23632 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSADGGQHW TYEGPHGQDH WPASYPECGN NAQSPIDIQT DSVTFDPDLP ALQPHGYDQP GTEPLDLHNN GHTVQLSLPS TLYLGGLPRK YVAAQLHLHW GQKGSPGGSE HQINSEATFA ELHIVHYDSD SYDSLSEAAE RPQGLAVLGI LIEVGETKNI AYEHILSHLH EVRHKDQKTS VPPFNLRELL PKQLGQYFRY NGSLTTPPCY QSVLWTVFYR RSQISMEQLE KLQGTLFSTE EEPSKLLVQN YRALQPLNQR MVFASFIQAG SSYTTGEM | |
Greater than 85.0% as determined by SDS-PAGE. |
CA14 Protein | |
Research Use Only | |
CA14 | |
Human | |
His | |
CA14 protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |