missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C7ORF49 Protein

A cDNA sequence encoding the C7ORF49 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11242-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15966588

  • £1320.00 / 100µg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

78996
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMETLQSE TKTRVLPSWL TAQVATKNVA PMKAPKRMRM AAVPVAAARL PATRTVYCMN EAEIVDVALG ILIESRKQEK ACEQPALAGA DNPEHSPPCS VSPHTSSGSS SEEEDSGKQA LAPGLSPSQR PGGSSSACSR SPEEEEEEDV LKYVREIFFS
Greater than 85% as determined by SDS-PAGE.
C7ORF49 Protein
Research Use Only
CYREN
Human
His
The C7ORF49 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers