missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C7ORF49 Protein

A cDNA sequence encoding the C7ORF49 was constructed and used to recombinantly synthesize the protein.

£104.00 - £1395.00

Specifications

Name C7ORF49 Protein
Product Type Recombinant Protein
Protein Tag His
Host Species E. coli
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15986588
View Documents Promotion Details
enQuireBio™
QP11242-2UG
2 μg
£104.00
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 20-09-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15976588
View Documents Promotion Details
enQuireBio™
QP11242-10UG
10 μg
£159.00
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 20-09-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15966588
View Documents Promotion Details
enQuireBio™
QP11242-100UG
100 μg
£1395.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 20-09-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

C7ORF49 Protein
His
Human
Greater than 85% as determined by SDS-PAGE.
CYREN
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMETLQSE TKTRVLPSWL TAQVATKNVA PMKAPKRMRM AAVPVAAARL PATRTVYCMN EAEIVDVALG ILIESRKQEK ACEQPALAGA DNPEHSPPCS VSPHTSSGSS SEEEDSGKQA LAPGLSPSQR PGGSSSACSR SPEEEEEEDV LKYVREIFFS
78996
SDS
Product Certifications