missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C1q Protein
A cDNA sequence encoding the C1q was constructed and used to recombinantly synthesize the protein.
£303.00 - £6015.00
Specifications
Name | C1q Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15966548
|
enQuireBio™
QP11228-200UG |
200 μg |
£303.00
200µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956548
|
enQuireBio™
QP11228-1MG |
1 mg |
£780.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946548
|
enQuireBio™
QP11228-10MG |
10 mg |
£6015.00
10mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
C1q Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 96.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Human | |
Lyophilized from 0.01M Na2 EDTA solution containing 0.3M NaCl pH 7.5. |