missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C17ORF49 Protein

A cDNA sequence encoding the C17ORF49 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11225-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15966538

  • £5741.17 / 1mg
Estimated Shipment: 07-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

C17ORF49 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The C17ORF49 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMTSASTK VGEIFSAAGA AFTKLGELTM QLHPVADSSP AGAKWTETEI EMLRAAVKRF GDDLNHISCV IKERTVAQIK ATVKRKVYED SGIPLPAESP KKGPKKVASG VLSPPPAAPP PSSSSVPEAG GPPIKKQKAD VTLSALNDSD ANSDVVDIEG LGETPPAKKL NFDQA
Videos
SDS
Documents
Certificates
Special Offers

Special Offers