missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C-C motif chemokine 25 Protein
A DNA sequence encoding the Homo sapiens (Human) C-C motif chemokine 25, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5774-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001188288.1 | |
C-C motif chemokine 25 Protein | |
Research Use Only | |
CCL25 | |
Recombinant Protein | |
E. coli | |
QGVFEDCCLA YHYPIGWAVL RRAWTYRIQE VSGSCNLPAA IFYLPKRHRK VCGNPKSREV QRAMKLLDAR NKVFAKLHHN TQTFQAGPHA VKKLSSGNSK LSSSKFSNPI SSSKRNVSLL ISANSGL | |
Greater than 90% as determined by SDS-PAGE. |
6370 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |