missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BTN3A2 Protein

A DNA sequence encoding the Homo sapiens (Human) BTN3A2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5730-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15967329

  • £1735.00 / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001184175.1
BTN3A2 Protein
Research Use Only
BTN3A2
Recombinant Protein
E. coli
QFSVLGPSGP ILAMVGEDAD LPCHLFPTMS AETMELKWVS SSLRQVVNVY ADGKEVEDRQ SAPYRGRTSI LRDGITAGKA ALRIHNVTAS DSGKYLCYFQ DGDFYEKALV ELKVAALGSN LHVEVKGYED GGIHLECRST GWYPQPQIQW SNAKGENIPA VEAPVVADGV GLYEVAASVI MRGGSGEGVS CIIRNSLLGL EKTASISIAD PFFRSAQPW
Greater than 90% as determined by SDS-PAGE.
11118
1 mg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers