missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BTD Protein
A cDNA sequence encoding the BTD was constructed and used to recombinantly synthesize the protein.
£99.15 - £1210.00
Specifications
Name | BTD Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15916488
|
enQuireBio™
QP11209-1UG |
1 μg |
£99.15
1µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936488
|
enQuireBio™
QP11209-5UG |
5 μg |
£155.00
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926488
|
enQuireBio™
QP11209-50UG |
50 μg |
£1210.00
50µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
BTD Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
BTD protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
AHTGEESVAD HHEAEYYVAA VYEHPSILSL NPLALISRQE ALELMNQNLD IYEQQVMTAA QKDVQIIVFP EDGIHGFNFT RTSIYPFLDF MPSPQVVRWN PCLEPHRFND TEVLQRLSCM AIRGDMFLVA NLGTKEPCHS SDPRCPKDGR YQFNTNVVFS NNGTLVDRYR KHNLYFEAAF DVPLKVDLIT FDTPFAGRFG IFTCFDILFF DPAIRVLRDY KVKHVVYPTA WMNQLPLLAA IEIQKAFAVA FGINVLAANV HHPVLGMTGS GIHTPLESFW YHDMENPKSH LIIAQVAKNP VGLIGAENAT GETDPSHSKF LKILSGDPYC EKDAQEVHCD EATKWNVNAP PTFHSEMMYD NFTLVPVWGK EGYLHVCSNG LCCYLLYERP TLSKELYALG VFDGLHTVHG TYYIQVCALV RCGGLGFDTC GQEITEATGI FEFHLWGNFS TSYIFPLFLT SGMTLEVPDQ LGWENDHYFL RKSRLSSGLV TAALYGRLYE RDLEHHHHHH | |
Greater than 85% as determined by SDS-PAGE. |