missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BRD3 Protein
A cDNA sequence encoding the BRD3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11204-1mg
Additional Details : Weight : 0.01000kg
Specifications
BRD3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSTATTV APAGIPATPG PVNPPPPEVS NPSKPGRKTN QLQYMQNVVV KTLWKHQFAW PFYQPVDAIK LNLPDYHKII KNPMDMGTIK KRLENNYYWS ASECMQDFNT MFTNCYIYNK PTDDIVLMAQ ALEKIFLQKV AQMPQEEVEL LPPAPKGKGR KPAAGAQSAG TQQVAAVSSV SPATPFQSVP PTVSQTPVIA ATPVPTITAN VTSVPVPPAA APPPPATPIV PVVPPTPPVV KKKGVKRKAD TTTPTTSAIT ASRSESPPPL SDPKQAKVVA RRESGGRPIK PPKKDLEDGE VPQHAGKKGK LSEHLRYCDS ILREMLSKKH AAYAWPFYKP VDAEALELHD YHDIIKHPMD LSTVKRKMDG REYPDAQGFA ADVRLMFSNC YKYNPPDHEV VAMARKLQDV FEMRFAKMP | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
BRD3 protein solution (0.5 mg/ml) containing PBS buffer (pH 7.4) and 10% glycerol. |