missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BP-1 Protein

A cDNA sequence encoding the BP-1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11197-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15976448

  • £302.00 / 5µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1748
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MTSLPCPLPG RDASKAVFPD LAPVPSVAAA YPLGLSPTTA ASPNLSYSRP YGHLLSYPYT EPANPGDSYL SCQQPAALSQ PLCGPAEHPQ ELEADSEKPR LSPEPSERRP QAPAKKLRKP RTIYSSLQLQ HLNQRFQHTQ YLALPERAQL AAQLGLTQTQ VKIWFQNKRS KYKKLLKQNS GGQEGDFPGR TFSVSPCSPP LPSLWDLPKA GTLPTSGYGN SFGAWYQHHS SDVLASPQMM
BP-1 Protein
Research Use Only
DLX4
Human
Untagged
BP-1 protein in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers