missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BMP10 Protein

A cDNA sequence encoding the BMP10 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10536-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15964168

  • £535.00 / 10µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

27302
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
The specific activity was determined by its ability to induce alkaline phosphatase production in the ATDC-5 cell line and is typically 4.0-6.0 ng/ml ED50.
Human
Untagged
The BMP10 protein was lyophilized from 10mM Sodium Citrate, pH 2.7 and 75mM NaCl .
BMP10 Protein
Research Use Only
BMP10
Recombinant Protein
Human (HEK293)
NAKGNYCKRT PLYIDFKEIG WDSWIIAPPG YEAYECRGVC NYPLAEHLTP TKHAIIQALVHLKNSQKASK ACCVPTKLEP ISILYLDKGV VTYKFKYEGM AVSECGCR
Greater than 95.0% as determined by analysis by SDS-PAGE and HPLC analysis.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers