missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BMP10 Protein
A cDNA sequence encoding the BMP10 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10536-5ug
Additional Details : Weight : 0.01000kg
Specifications
27302 | |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The specific activity was determined by its ability to induce alkaline phosphatase production in the ATDC-5 cell line and is typically 4.0-6.0 ng/ml ED50. | |
Human | |
Untagged | |
The BMP10 protein was lyophilized from 10mM Sodium Citrate, pH 2.7 and 75mM NaCl . |
BMP10 Protein | |
Research Use Only | |
BMP10 | |
Recombinant Protein | |
Human (HEK293) | |
NAKGNYCKRT PLYIDFKEIG WDSWIIAPPG YEAYECRGVC NYPLAEHLTP TKHAIIQALVHLKNSQKASK ACCVPTKLEP ISILYLDKGV VTYKFKYEGM AVSECGCR | |
Greater than 95.0% as determined by analysis by SDS-PAGE and HPLC analysis. |