missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BDNF Protein

Recombinant Protein

Brand:  enQuireBio™ QP5220-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15965729

  • £3285.00 / 500µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001137277.1
BDNF Protein
Research Use Only
BDNF
Recombinant Protein
E. coli
MHSDPARRGE LSVCDSISEW VTAADKKTAV DMSGGTVTVL EKVPVSKGQL KQYFYETKCN PMGYTKEGCR GIDKRHWNSQ CRTTQSYVRA LTMDSKKRIG WRFIRIDTSC VCTLTIKRGR
Greater than 95% as determined by SDS-PAGE.
627
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
To pass QC, Human BDNF must induce C6 cell proliferation significantly at concentrations less than 2 ug/ml, or activity of at least 5.0 x 10^2 units/mg (typical ED50 is less than 1 ug/ml).
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers