missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BDNF Protein
Recombinant Protein
Brand: enQuireBio™ QP5220-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001137277.1 | |
BDNF Protein | |
Research Use Only | |
BDNF | |
Recombinant Protein | |
E. coli | |
MHSDPARRGE LSVCDSISEW VTAADKKTAV DMSGGTVTVL EKVPVSKGQL KQYFYETKCN PMGYTKEGCR GIDKRHWNSQ CRTTQSYVRA LTMDSKKRIG WRFIRIDTSC VCTLTIKRGR | |
Greater than 95% as determined by SDS-PAGE. |
627 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
To pass QC, Human BDNF must induce C6 cell proliferation significantly at concentrations less than 2 ug/ml, or activity of at least 5.0 x 10^2 units/mg (typical ED50 is less than 1 ug/ml). | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |