missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BCOADC-E2 Protein
A cDNA sequence encoding the BCOADC-E2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11148-1mg
Additional Details : Weight : 0.01000kg
Specifications
BCOADC-E2 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
DLST (0.91 mg/ml) is supplied in 20mM HEPES buffer pH 8.0, 200mM NaCl and 20% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Greater than 75% as determined by SDS-PAGE. |