missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BCOADC-E2 Protein
A cDNA sequence encoding the BCOADC-E2 was constructed and used to recombinantly synthesize the protein.
£99.15 - £4515.00
Specifications
Name | BCOADC-E2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976298
|
enQuireBio™
QP11148-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956298
|
enQuireBio™
QP11148-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966298
|
enQuireBio™
QP11148-1MG |
1 mg |
£4515.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
BCOADC-E2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
Greater than 75% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
DLST (0.91 mg/ml) is supplied in 20mM HEPES buffer pH 8.0, 200mM NaCl and 20% glycerol. |