missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BATF Protein

A cDNA sequence encoding the BATF was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11128-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956238

  • £155.00 / 20µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

BATF Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MPHSSDSSDS SFSRSPPPGK QDSSDDVRRV QRREKNRIAA QKSRQRQTQK ADTLHLESED LEKQNAALRK EIKQLTEELK YFTSVLNSHE PLCSVLAAST PSPPEVVYSA HAFHQPHVSS PRFQP
BATF purity was found to be greater than 90.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The BATF solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 2mM DTT and 40% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers