missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Baculoviral IAP repeat-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Baculoviral IAP repeat-containing protein 1, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001333799.1 |
---|---|
Gene ID (Entrez) | 4671 |
Name | Baculoviral IAP repeat-containing protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16069310
|
enQuireBio™
QP7547-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16089310
|
enQuireBio™
QP7547-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16079310
|
enQuireBio™
QP7547-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099310
|
enQuireBio™
QP7547-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009320
|
enQuireBio™
QP7547-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16019320
|
enQuireBio™
QP7547-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001333799.1 | |
Baculoviral IAP repeat-containing protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
4671 | |
Research Use Only | |
NAIP | |
Recombinant Protein | |
E. coli | |
EAKRLKTFVT YEPYSSWIPQ EMAAAGFYFT GVKSGIQCFC CSLILFGAGL TRLPIEDHKR FHPDCGFLLN KDVGNIAKYD IRVKNLKSRL RGGKMRYQEE EARLASFRNW PFYVQGISPC VLSEAGFVFT GKQDTVQCFS CGGCLGNWEE GDDPWKEHAK WFPKCEFLRS KKSSEEITQY IQSYKGFVDI TGEHFVNSWV QRELPMASAY CNDSIFAYEE LRLDSFKDWP RESAVGVAAL AKAGLFYTGI KDIVQCFSCG GCLEKWQEGD DPLDDHTRCF PNCPFL | |
Greater than 90% as determined by SDS-PAGE. |