missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BAALC Protein

A cDNA sequence encoding the BAALC was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11121-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956218

  • £99.15 / 2µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

BAALC Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGCGGSRADA IEPRYYESWT RETESTWLTY TDSDAPPSAA APDSGPEAGG LHSGMLEDGL PSNGVPRSTA PGGIPNPEKK TNCETQCPNP QSLSSGPLTQ KQNGLQTTEA KRDAKRMPAK EVTINVTDSI QQMDRSRRIT KNCVN
Greater than 95.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The protein solution contains 20mM Tris buffer pH 8.0, 1mM DTT, 20% glycerol and 2mM EDTA.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers