missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AZGP1 Protein

A cDNA sequence encoding the AZGP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11114-HIS-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15966188

  • £3973.41 / 1mg
Estimated Shipment: 17-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

563
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
AZGP1
Human
His
AZGP1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol.
AZGP1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSQENQDGR YSLTYIYTGL SKHVEDVPAF QALGSLNDLQ FFRYNSKDRK SQPMGLWRQV EGMEDWKQDS QLQKAREDIF METLKDIVEY YNDSNGSHVL QGRFGCEIEN NRSSGAFWKY YYDGKDYIEF NKEIPAWVPF DPAAQITKQK WEAEPVYVQR AKAYLEEECP ATLRKYLKYS KNILDRQDPP SVVVTSHQAP GEKKKLKCLA YDFYPGKIDV HWTRAGEVQE PELRGDVLHN GNGTYQSWVV VAVPPQDTAP YSCHVQHSSL AQPLVVPWEA S
Videos
SDS
Documents
Certificates
Special Offers

Special Offers