missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATP5D Protein
A DNA sequence encoding the Homo sapiens (Human) ATP5D, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5695-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001001975.1 | |
ATP5D Protein | |
Research Use Only | |
ATP5D | |
Recombinant Protein | |
Yeast | |
AEAAAAPAAA SGPNQMSFTF ASPTQVFFNG ANVRQVDVPT LTGAFGILAA HVPTLQVLRP GLVVVHAEDG TTSKYFVSSG SIAVNADSSV QLLAEEAVTL DMLDLGAAKA NLEKAQAELV GTADEATRAE IQIRIEANEA LVKALE | |
Greater than 90% as determined by SDS-PAGE. |
513 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |