missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ATP5D Protein

A DNA sequence encoding the Homo sapiens (Human) ATP5D, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5695-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15967069

  • £384.00 / 100µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001001975.1
ATP5D Protein
Research Use Only
ATP5D
Recombinant Protein
E. coli
AEAAAAPAAA SGPNQMSFTF ASPTQVFFNG ANVRQVDVPT LTGAFGILAA HVPTLQVLRP GLVVVHAEDG TTSKYFVSSG SIAVNADSSV QLLAEEAVTL DMLDLGAAKA NLEKAQAELV GTADEATRAE IQIRIEANEA LVKALE
Greater than 90% as determined by SDS-PAGE.
513
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers