missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATOX1 Protein
A cDNA sequence encoding the ATOX1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11103-1mg
Additional Details : Weight : 0.01000kg
Specifications
ATOX1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPKHEFSVDM TCGGCAEAVS RVLNKLGGVK YDIDLPNKKV CIESEHSMDT LLATLKKTGK TVSYLGLE | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ATOX1 solution containing 20mM Tris pH 8.0, 1mM DTT and 10% glycerol. |