missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATF4 Protein
A cDNA sequence encoding the ATF4 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Gene ID (Entrez) | 468 |
---|---|
Name | ATF4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | ATF4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15946138
|
enQuireBio™
QP11099-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926138
|
enQuireBio™
QP11099-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936138
|
enQuireBio™
QP11099-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
468 | |
Research Use Only | |
ATF4 | |
Human | |
Untagged | |
ATF4 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol. |
ATF4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MHHHHHHMAD QLTEEQIAEF KEAFSLFDKD GDGTITTKEL GTVMRSLGQN PTEAELQDMI NEVDADGNGT IDFPEFLTMM ARKMKDTDSE EEIREAFRVF DKDGNGYISA AELRHVMTNL GEKLTDEEVD EMIREADIDG DGQVNYEEFV QMMTAKGSHM TEMSFLSSEV LVGDLMSPFD QSGLGAEESL GLLDDYLEVA KHFKPHGFSS DKAKAGSSEW LAVDGLVSPS NNSKEDAFSG TDWMLEKMDL KEFDLDALLG IDDLETMPDD LLTTLDDTCD LFAPLVQETN KQPPQTVNPI GHLPESLTKP DQVAPFTFLQ PLPLSPGVLS STPDHSFSLE LGSEVDITEG DRKPDYTAYV AMIPQCIKEE DTPSDNDSGI CMSPESYLGS PQHSPSTRGS PNRSLPSPGV LCGSARPKPY DPPGEKMVAA KVKGEKLDKK LKKMEQNKTA ATRYRQKKRA EQEALTGECK ELEKKNEALK ERADSLAKEI QYLKDLIEEV RKARGKKRVP | |
Greater than 90.0% as determined by SDS-PAGE. |