missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ASRGL1 Protein
A cDNA sequence encoding the ASRGL1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11093-1mg
Additional Details : Weight : 0.01000kg
Specifications
80150 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMNPIVVV HGGGAGPISK DRKERVHQGM VRAATVGYGI LREGGSAVDA VEGAVVALED DPEFNAGCGS VLNTNGEVEM DASIMDGKDL SAGAVSAVQC IANPIKLARL VMEKTPHCFL TDQGAAQFAA AMGVPEIPGE KLVTERNKKR LEKEKHEKGA QKTDCQKNLG TVGAVALDCK GNVAYATSTG GIVNKMVGRV GDSPCLGAGG YADNDIGAVS TTGHGESILK VNLARLTLFH IEQGKTVEEA ADLSLGYMKS RVKGLGGLIV VSKTGDWVAK WTSTSMPWAA AKDGKLHFGI DPDDTTITDL P | |
Greater than 90.0% as determined by SDS-PAGE. |
ASRGL1 Protein | |
Research Use Only | |
ASRGL1 | |
Human | |
His | |
ASRGL1 protein solution (0.5 mg/ml) containing Phosphate buffer saline, (pH 7.4) ,10% glycerol and 1mM DTT. |