missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ASPHD1 Protein
A cDNA sequence encoding the ASPHD1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | ASPHD1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976108
|
enQuireBio™
QP11090-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966108
|
enQuireBio™
QP11090-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956108
|
enQuireBio™
QP11090-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ASPHD1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ASPHD1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSYRLGSQD MQALGAGSRA GGVRGGPVGC SEAGGPSPGG PGDPGEGPRT EGLVSRRLRA YARRYSWAGM GRVRRAAQGG PGPGRGPGVL GIQRPGLLFL PDLPSAPFVP RDAQRHDVEL LESSFPAILR DFGAVSWDFS GTTPPPRGWS PPLAPGCYQL LLYQAGRCQP SNCRRCPGAY RALRGLRSFM SANTFGNAGF SVLLPGARLE GRCGPTNARV RCHLGLKIPP GCELVVGGEP QCWAEGHCLL VDDSFLHTVA HNGSPEDGPR VVFIVDLWHP NVAGAERQAL DFVFAPDP | |
Greater than 90.0% as determined by SDS-PAGE. |