missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ART1 / CD296 / ARTC1 Protein

A DNA sequence encoding the Homo sapiens (Human) ART1 / CD296 / ARTC1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5681-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15946959

  • £1735.33 / 1mg
Estimated Shipment: 17-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004305.2
ART1 / CD296 / ARTC1 Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
417
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ART1
Recombinant Protein
E. coli
QSHPITRRDL FSQEIQLDMA LASFDDQYAG CAAAMTAALP DLNHTEFQAN QVYADSWTLA SSQWQERQAR WPEWSLSPTR PSPPPLGFRD EHGVALLAYT ANSPLHKEFN AAVREAGRSR AHYLHHFSFK TLHFLLTEAL QLLGSGQRPP RCHQVFRGVH GLRFRPAGPR ATVRLGGFAS ASLKHVAAQQ FGEDTFFGIW TCLGAPIKGY SFFPGEEEVL IPPFETFQVI NASRLAQGPA RIYLRALGKH STYNCEYIKD KKCKSGPCHL DNS
Videos
SDS
Documents
Certificates
Special Offers

Special Offers