missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ARL15 Protein
A cDNA sequence encoding the ARL15 was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | ARL15 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976048
|
enQuireBio™
QP11070-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966048
|
enQuireBio™
QP11070-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956048
|
enQuireBio™
QP11070-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ARL15 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ARL15 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSDLRITEAF LYMDYLCFRA LCCKGPPPAR PEYDLVCIGL TGSGKTSLLS KLCSESPDNV VSTTGFSIKA VPFQNAILNV KELGGADNIR KYWSRYYQGS QGVIFVLDSA SSEDDLEAAR NELHSALQHP QLCTLPFLIL ANHQDKPAAR SVQEIKKYFE LEPLARGKRW ILQPCSLDDM DALKDSFSQL INLLEEKDHE AVRM | |
Greater than 85% as determined by SDS-PAGE. |